Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHMT2 Antibody (OTI3E9), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274171AF700
Description
SHMT2 Monoclonal specifically detects SHMT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SHMT2 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SHMT2 | |
Full length recombinant protein of human SHMT2 (NP_005403) produced in E.coli. | |
0.1 mL | |
6472.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3E9 | |
Western Blot, Immunohistochemistry | |
EC 2.1.2.1, GLY A+, GLYA, glycine auxotroph A, human complement for hamster, Glycine hydroxymethyltransferase, serine aldolase, serine hydroxymethylase, serine hydroxymethyltransferase 2 (mitochondrial), serine hydroxymethyltransferase, mitochondrial, Serine methylase, SHMT, threonine aldolase | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction