Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHMT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542822
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542822 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542822 Supplier Invitrogen™ Supplier No. PA542822
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ELIASENFCS RAALEALGSC LNNKYSEGYP GKRYYGGAEV VDEIELLCQR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Mammalian serine hydroxymethyltransferase (SHMT) is a tetrameric, pyridoxal phosphate (PLP)-dependent enzyme that catalyzes the reversible interconversion of serine and tetrahydrofolate to glycine and methylenetetrahydrofolate in the cytoplasm (cSHMT, SHMT1) and mitochondria (mSHMT, SHMT2). cSHMT preferentially supplies one-carbon units for thymidylate biosynthesis, depletes methylenetetrahydrofolate pools for S-adenosylmethionine (SAM) synthesis by synthesizing serine, sequesters 5-methyltetrahydrofolate, and inhibits SAM synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHMT2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SHMT2
Gene Accession No. P34897
Gene Alias 2700043D08Rik; AA408223; AA986903; epididymis secretory sperm binding protein Li 51e; GLY A+; GLYA; glycine auxotroph A, human complement for hamster; glycine hydroxymethyltransferase; HEL-S-51e; mitochondrial serine hydroxymethyltransferase; serine aldolase; serine hydroxymethylase; serine hydroxymethyltransferase; serine hydroxymethyltransferase 2; serine hydroxymethyltransferase 2 (mitochondrial); serine hydroxymethyltransferase, mitochondrial; Serine methylase; SHMT; SHMT 2; SHMT2; threonine aldolase
Gene Symbols SHMT2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SHMT2 (aa 72-121).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6472
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.