Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SHMT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554232
Description
Immunogen sequence: NTCPGDRSAI TPGGLRLGAP ALTSRQFRED DFRRVVDFID EGVNIGLEVK SKTAKLQDFK SFLLKDSETS QRLANLRQRV EQFARAF Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Mammalian serine hydroxymethyltransferase (SHMT) is a tetrameric, pyridoxal phosphate (PLP)-dependent enzyme that catalyzes the reversible interconversion of serine and tetrahydrofolate to glycine and methylenetetrahydrofolate in the cytoplasm (cSHMT, SHMT1) and mitochondria (mSHMT, SHMT2). cSHMT preferentially supplies one-carbon units for thymidylate biosynthesis, depletes methylenetetrahydrofolate pools for S-adenosylmethionine (SAM) synthesis by synthesizing serine, sequesters 5-methyltetrahydrofolate, and inhibits SAM synthesis.
Specifications
| SHMT2 | |
| Polyclonal | |
| Unconjugated | |
| SHMT2 | |
| 2700043D08Rik; AA408223; AA986903; epididymis secretory sperm binding protein Li 51e; GLY A+; GLYA; glycine auxotroph A, human complement for hamster; glycine hydroxymethyltransferase; HEL-S-51e; mitochondrial serine hydroxymethyltransferase; serine aldolase; serine hydroxymethylase; serine hydroxymethyltransferase; serine hydroxymethyltransferase 2; serine hydroxymethyltransferase 2 (mitochondrial); serine hydroxymethyltransferase, mitochondrial; Serine methylase; SHMT; SHMT 2; SHMT2; threonine aldolase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 108037, 299857, 6472 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P34897, Q9CZN7 | |
| SHMT2 | |
| Recombinant protein corresponding to Human SHMT2. Recombinant protein control fragment (Product #RP-91763). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction