Learn More
Invitrogen™ SHMT2 Polyclonal Antibody

Description
Recommended positive controls: 293T, HeLa, A431, HepG2. Predicted reactivity: Mouse (93%), Rat (91%), Xenopus laevis (81%), Rabbit (93%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | SHMT2 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | SHMT2 |
| Gene Accession No. | P34897, Q9CZN7 |
| Gene Alias | 2700043D08Rik; AA408223; AA986903; epididymis secretory sperm binding protein Li 51e; GLY A+; GLYA; glycine auxotroph A, human complement for hamster; glycine hydroxymethyltransferase; HEL-S-51e; mitochondrial serine hydroxymethyltransferase; serine aldolase; serine hydroxymethylase; serine hydroxymethyltransferase; serine hydroxymethyltransferase 2; serine hydroxymethyltransferase 2 (mitochondrial); serine hydroxymethyltransferase, mitochondrial; Serine methylase; SHMT; SHMT 2; SHMT2; threonine aldolase |
| Gene Symbols | SHMT2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.