Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHOC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578243
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578243 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578243 Supplier Invitrogen™ Supplier No. PA578243
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: A431 Predicted Reactivity: Rat (100%), Zebrafish (97%), Xenopus laevis (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SHOC2 protein participates in protein binding / transferase activity in the fibroblast growth factor receptor signaling pathway and Ras protein signal transduction. It is a widely expressed protein composed almost entirely of leucine-rich repeats (LRR), with a lysine-rich sequence at the amino terminus and cytoplasmically localized. SHOC2 acts as a positive modulator of the RAS-MAPK signaling cascade, which is elicited by EGL-15 and LET-23 and mediated by LET-60. SHOC2 together with protein phosphatase 1c (PP1c) forms a highly specific M-Ras effector complex and is essential for activation of the MAPK pathway by growth factors. Furthermore, in tumor cells with Ras gene mutations, inhibition of SHOC2 expression inhibits MAPK, but not PI3K activity. The SHOC2-PP1c holoenzyme provides an attractive therapeutic target for inhibition of the MAPK pathway in cancer. Recent studies show that aberrantly acquired N-myristoylation of SHOC2 causes human disease Noonan-like syndrome with loose anagen hair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHOC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Shoc2
Gene Accession No. O88520, Q9UQ13
Gene Alias AU017287; KIAA0862; Leucine-rich repeat protein SHOC-2; mKIAA0862; Protein soc-2 homolog; protein sur-8 homolog; Ras-binding protein SUR-8; RP11-348N5.2; Shoc2; SHOC2 leucine-rich repeat scaffold protein; SHOC2, leucine rich repeat scaffold protein; SIAA0862; SOC2; soc-2 (suppressor of clear) homolog; soc-2 (suppressor of clear) homolog (C. elegans); soc-2 suppressor of clear homolog; soc-2 suppressor of clear homolog (C. elegans); suppressor of clear, C. elegans, homolog of; SUR8
Gene Symbols Shoc2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human SHOC2. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56392, 8036
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.