Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOTRAX™ Shoe Sanitizer System
Shop All NoTrax Products
Click to view available options
Dimensions (L x W):
52.5 x 25.5 in.
Description
- Simple, safe and effective shoe disinfecting system to limit the spread of contaminants on floors
- Labels clearly instruct how to sanitize shoes
- Ideal for entrances to schools, businesses, facilities, and more
- Low profile bevels for easy walk on/off to minimize trip hazards
- Designed as a zoned system with completely contained and secured components
- Carpet mat inserts are disposable and replaceable for quick and easy cleaning
- Available in 2-zone (disinfecting, drying) and 3-zone (cleaning, disinfecting, drying) options
- Each system cleans and sanitizes shoe soles, then quickly dries them the moment you step out
- Mat inserts are adhesive backed and replaceable for easy cleaning
- Carpet mats are made from recycled plastic bottles
- System is lightweight and easy to set up and move
- Vinyl backed to prevent slipping
Specifications
Specifications
Length (Metric) | 133.4 cm |
Type | Entrance Mat |
Width (English) | 25.5 in. |
Description | Shoe Disinfecting and Drying System (2-Zone) |
Width (Metric) | 64.8 cm |
Dimensions (L x W) | 52.5 x 25.5 in. |
Material | Recycled Plastic |
Color | Black |
Length (English) | 52.5 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction