Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Short-Term Detector Tubes - Perchloroethylene

Catalog No. 17985283 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Components Measured:
Perchloroethylene 0.1/a
Perchloroethylene 2/a
Measurement Range:
0.5 to 4ppm
2 to 300ppm
Humidity Range:
30mg/L
Less than25mg/L
No. of Strokes:
1/5
3/9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Measurement Range Humidity Range No. of Strokes
17-985-283 Perchloroethylene 2/a 2 to 300ppm Less than25mg/L 1/5
19-046-631 Perchloroethylene 0.1/a 0.5 to 4ppm 30mg/L 3/9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-985-283 Supplier Dräger™ Supplier No. 8101501
Add to Cart
Add to Cart

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Components Measured Perchloroethylene 2/a
Humidity Range Less than25mg/L
Measurement Range 2 to 300ppm
No. of Strokes 1/5
Quantity 10
Response Time 3 min./30 sec.
Temperature Range (English) 59°F to 86°F
Temperature Range (Metric) 15°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.