Learn More
Dräger™ Short-Term Detector Tubes: Toluene
Catalog No. 18556K Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
100 to 1800
5–300ppm
50 to 400
Tube Type:
Toluene 100/a
Toluene 5/b
Toluene 50/a
Humidity Range:
5 to 12mg/L
Less than30mg/L
Not Applicable
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Tube Type Humidity Range
18-556K 50 to 400 Toluene 50/a 5 to 12mg/L
18-556J 5–300ppm Toluene 5/b Not Applicable
18-556L 100 to 1800 Toluene 100/a Less than30mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-556K Supplier Dräger™ Supplier No. 8101701
Add to Cart
Add to Cart
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Components Measured Toluene
Humidity Range 5 to 12mg/L
Measurement Range 50 to 400
No. of Strokes 5
Quantity 10
Response Time 1.5 min.
Temperature Range (English) 32°F to 86°F
Temperature Range (Metric) 0°C to 30°C
Tube Type Toluene 50/a