Description
- ANSI cut level A3
- ISO 13997 cut level C
- Cut-resistant liner
- Ergonomic design for maximum comfort
- Unparalleled grip
- Sponge nitrile technology absorbs oil and water
- 15-gauge seamless stretch, cut-resistant liner
- Made with Kevlar™ Aramid fibers
- Color coded cuff
- Maximum Oil Absorbency Sponge nitrile coated palm
Specifications
Specifications
| Certifications/Compliance | ISO 13997 |
| Color | Yellow |
| Cut Resistance Rating | ANSI A3 |
| Product Type | Cut-resistant Gloves |
| Size | 2X-Large |
| Glove Cuff Color | Gray |
| Liner Material | Aramid Fiber |