Learn More
SHOWA™ 541 HPPE Cut-Resistant Gloves, Polyurethane Coating
Catalog No. 19270224
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Rough grip gloves provide a high degree of comfort and control when performing intricate tasks. Dark color hides grime in dirty environments.
  • Durable flat-dipped gray colored polyurethane-coated glove
  • 13-gauge HPPE cut-resistant liner
  • Elastic knit-wrist secures glove above wrist for comfort and prevents debris from entering glove
Specifications
Certifications/Compliance (Europe) Cat II CE
Construction Seamless Knit
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A3
Material HPPE
Product Type Cut-resistant Gloves
Size X-Large
EN388 Standard EN 388:4342B
For Use With (Application) Intricate sharp parts handling, construction, glass handling, bottling, canning, sheet metal work, automotive, fabrication
Glove Cuff Style Elasticated
Model 541-XL
Puncture Rating ANSI/ISEA 105-16 Level 3
Series 541 Series
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.