Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 545 Nitrile Coated Seamless Knit HPPE Gloves

Catalog No. 19270231 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-270-231 Medium
19-270-232 Large
19-270-233 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-270-231 Supplier SHOWA™ Supplier No. 545M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Nitrile coating on the palm enhances grip capabilities, in addition to providing cut and oil resistance. The elasticated wrist cuff prevents debris from entering the glove to ensure comfort over long periods of wear.

  • Nitrile coating enhances grip capabilities, as well as oil and cut resistance
  • Elasticated wrist cuff prevents debris from entering the glove
  • A high level of precision and dexterity, even when assembling greasy components
  • Ensures comfort over long periods of wear

Specifications

Construction Seamless Knit
Coating Material Nitrile
Color Gray
Cut Resistance Rating ANSI A2
Exterior Finish Fully Textured
Hand Specific
Material HPPE
Product Type Seamless Knit Gloves
Size Medium
EN388 Standard 4242B
For Use With (Application) Manufacturing, Machine Operation, and Mining
Glove Cuff Style Knit
Series 545
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.