Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ AEGIS KVS4™ Cut-Resistant Gloves

Catalog No. 19120645 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
2X-Large
Length (English):
7 in.
11 in.
Length (Metric):
17.8 cm
27.9 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (English) Length (Metric)
19-120-645 2X-Large 11 in. 27.9 cm
19-120-641 Small 7 in. 17.8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-120-645 Supplier SHOWA™ Supplier No. 25011
This item is not returnable. View return policy

Thin, lightweight seamless knit gloves are comprised of HPPE, nylon, lycra with Zorb-IT™ palm coating for oily grip.

  • 10-gauge seamless knit comprised
  • High Performance Poly Ethylene (HPPE), nylon, lycra with Zorb-IT™ palm coating
  • Combination of comfort, cut resistance and durability
  • Thin, lightweight fit and feel

Specifications

Coating Material Sponge Nitrile
Color Black, Yellow
Cut Resistance Rating ANSI A4
Material Aramid Fiber, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size 2X-Large
For Use With (Application) Oily surfaces, Metal stamping, mechanical and automotive, sheet Metal assembly, Glass handling, bottling, recycling
Length (English) 11 in.
Length (Metric) 27.9 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.