Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 377 Nitrile Foam Coated Chemical Resistant Gloves

Catalog No. 19020701 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-020-701 2X-Large
19-020-700 Small
19-172-208 Medium
19-172-209 Large
19-172-210 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-020-701 Supplier SHOWA™ Supplier No. 377XXL10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves feature a nitrile foam coating for enhanced grip capabilities in slippery environments, with a seamless liner and elasticated cuff to keep hands warm and protected.

  • Chemical resistant gloves with ultra-tactile grip and durability
  • Nitrile foam coating enhances grip capabilities in slippery environments
  • Seamless liner and elasticated cuff to keep hands warm and protected

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile Foam
Color Blue, Black
Hand Specific
Latex Free Yes
Material Nylon, Polyester
Product Type Dip-coated Gloves
Size 2X-Large
EN388 Standard 4121X
For Use With (Application) Agriculture, Construction, and Manufacturing
Glove Cuff Color Gray
Glove Cuff Style Elasticated
Series 377
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.