Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Polyurethane Coated Seamless Knit Nylon Gloves

Catalog No. 191304682A Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-4682A Small
19-130-4682B Medium
19-130-4682C Large
19-130-4682D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-4682A Supplier SHOWA™ Supplier No. BO500WS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves are designed for precision and cleanliness with a permeable nitrile coating, breathable back, and seamless liner to prevent irritation.

  • Snug fit maximizes tactile feedback
  • Coating is permeable, allowing hands to breathe to reduce perspiration
  • Light, elastic low-lint glove is resistant to deformation
  • Polyurethane protects the hand from oils and abrasion
  • Seamless construction eliminates irritation
  • White color is ideal for clean room applications
  • Mimimizes particulates
  • No latex allergy risks

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Cuff Material Elastic
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
EN388 Standard 3131X
For Use With (Application) Intricate Parts Handling, Clean Rooms, Automotive Repairs, and Integrated Circuits
Glove Cuff Color Natural
Glove Cuff Style Knit
Series BO500W
Yarn Gauge 13
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.