Learn More
Description
- Outstanding ratio of cut resistance to weight
- Good touch and tactile sensitivity
- Suits assembly workers handling jagged rivets and sharp parts, as well as machinists replacing pinions and sharp-edged tooling parts
- Ambidextrous
Specifications
Specifications
| Color | Light Gray |
| Cut Resistance Rating | ANSI A3 |
| Hand | Ambidextrous |
| Material | Dyneema™, AlphaSan |
| Product Type | Cut-resistant Gloves |
| Size | X-Large, 10 |
| For Use With (Application) | Meat cutting, construction, glove liner, recycling, bottling, glass handling, sheet metal handling, razor blades |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.