Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ T-Flex™ Cut-Resistant Gloves

Catalog No. 191301055 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Large, 9
Medium, 8
Small, 7
X-Large, 10
X-Small, 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-1055 Small, 7
19-169-950 X-Small, 6
19-130-1056 Medium, 8
19-130-1057 Large, 9
19-130-1054 X-Large, 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-130-1055 Supplier SHOWA™ Supplier No. 811507
This item is not returnable. View return policy

Thin, lightweight glove boasts a construction made up of only Dyneema™ cut-resistant fibers

  • Outstanding ratio of cut resistance to weight
  • Good touch and tactile sensitivity
  • Suits assembly workers handling jagged rivets and sharp parts, as well as machinists replacing pinions and sharp-edged tooling parts
  • Ambidextrous

Specifications

Color Light Gray
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Material Dyneema™, AlphaSan
Product Type Cut-resistant Gloves
Size Small, 7
For Use With (Application) Meat cutting, construction, glove liner, recycling, bottling, glass handling, sheet metal handling, razor blades
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.