Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOX2 Mouse anti-Human, Rat, Unconjugated, Clone: 1D1, Abnova™

Catalog No. 89339157 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-157 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-157 Supplier Abnova Corporation Supplier No. H00006474M01J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

SHOX2 Monoclonal antibody specifically detects SHOX2 in Human, Rat samples. It is validated for WB-Ce, S-ELISA, ELISA, WB-Re, WB-Tr, RNAi-Ab.

Specifications

Antigen SHOX2
Applications ELISA, Western Blot, KnockDown
Classification Monoclonal
Clone 1D1
Conjugate Unconjugated
Description short stature homeobox 2
Gene SHOX2
Gene Accession No. NM_006884
Gene Alias OG12, OG12X, OGI2X, SHOT
Host Species Mouse
Immunogen SHOX2 (NP_006875, 117 a.a. ∼ 204 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6474
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.