Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHOX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596999
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596999 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596999 Supplier Invitrogen™ Supplier No. PA596999
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELDMGAAERS REPGSPRLTE GRRKPTKAEV QATLLLPGEA FRFLVSPELK DRKEDAKGME DEGQTKIKQR RSRTNFTLEQ LNELERLFDE THYPDAFMRE ELSQRLGLSE ARVQVWFQNR RAKCRKQENQ LHKGVLIGAA SQFEACRVAP YVNVGALRMP FQQDSHCNVT PLSFQVQAQL QLDSAVAHAH HHLHPHLAAH APYMMFPAPP FGLPLATLAA DSASAASVVA AAAAAKTTSK NSSIADLRLK AKKHAAALGL; Positive Samples: NCI-H460, Mouse skeletal muscle, Mouse liver; Cellular Location: Nucleus.

SHOX2 is a member of the homeo box family of genes that encode proteins containing a 60-amino acid residue motif that represents a DNA binding domain. Homeo box genes have been characterized extensively as transcriptional regulators involved in pattern formation in both invertebrate and vertebrate species. Several human genetic disorders are caused by aberrations in human homeo box genes. SHOX is a pseudoautosomal homeo box gene that is thought to be responsible for idiopathic short stature and implicated to play a role in the short stature phenotype of Turner syndrome patients. This gene is a member of the homeo box family of genes that encode proteins containing a 60-amino acid residue motif that represents a DNA binding domain. Homeo box genes have been characterized extensively as transcriptional regulators involved in pattern formation in both invertebrate and vertebrate species. Several human genetic disorders are caused by aberrations in human homeo box genes. SHOX is a pseudoautosomal homeo box gene that is thought to be responsible for idiopathic short stature and implicated to play a role in the short stature phenotype of Turner syndrome patients. This gene is considered to be a candidate gene for Cornelia de Lange syndrome. Alternative splicing has been observed at this locus and two variants, each encoding a distinct isoform, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHOX2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SHOX2
Gene Accession No. O35750, O60902, P70390
Gene Alias 6330543G17Rik; Homeobox protein Og12X; OG12; OG-12; Og12x; OGI2X; Paired family homeodomain protein Prx3; paired-related homeobox protein SHOT; Prx3; short stature homeobox 2; short stature homeobox protein 2; SHOT; SHOX homologous gene on chromosome 3; Shox2; similar to short stature homeobox 2; zgc:65884; zgc:77344
Gene Symbols SHOX2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 96-355 of human SHOX2 (NP_0030213).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20429, 25546, 6474
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.