Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHP-1 Antibody (CPTC-PTPN6-2), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308900AF750
Description
SHP-1 Monoclonal antibody specifically detects SHP-1 in Human samples. It is validated for Western Blot, ELISASpecifications
SHP-1 | |
Monoclonal | |
Alexa Fluor 750 | |
HCP, HCPH, HPTP1C, PTP-1C, PTPN6, SHP1, SHP-1L, SH-PTP1 | |
Recombinant human full-length SHP-1 protein (Uniprot: P29350) | |
100 μg | |
Apoptosis, Protein Phosphatase | |
5777 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA | |
CPTC-PTPN6-2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction