Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHP-1 Rabbit anti-Human, Mouse, Rat, Clone: 7I1K3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | SHP-1 |
---|---|
Clone | 7I1K3 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
SHP-1 Monoclonal antibody specifically detects SHP-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
SHP-1 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis, Protein Phosphatase | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5777 | |
IgG | |
Affinity purified |
7I1K3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
HCP, HCPH, HPTP1C, PTP-1C, PTPN6, SHP1, SHP-1L, SH-PTP1 | |
A synthetic peptide corresponding to a sequence within amino acids 496-595 of human SHP-1 (P29350). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title