Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHP-2/PTPN11 Antibody (OTI3F2), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274173B
Description
SHP-2/PTPN11 Monoclonal specifically detects SHP-2/PTPN11 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry.Specifications
SHP-2/PTPN11 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
PTPN11 | |
Full length recombinant protein of human SHP-2/PTPN11 (NP_002825) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG2a |
Western Blot, Flow Cytometry | |
OTI3F2 | |
Western Blot, Flow Cytometry | |
BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP-1D, PTP-2C, PTP2CNS1, Shp2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11 | |
Mouse | |
Affinity Purified | |
Angiogenesis, Cancer, Cell Biology, Lipid and Metabolism, Phospho Specific, Signal Transduction, Tyrosine Kinases | |
5781.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction