Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHP-2/PTPN11, p Tyr542 Antibody (2E2), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | SHP-2/PTPN11 |
---|---|
Clone | 2E2 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000 |
Applications | Western Blot, ELISA, Immunoprecipitation |
Classification | Monoclonal |
Description
SHP-2/PTPN11 Monoclonal antibody specifically detects SHP-2/PTPN11 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
SHP-2/PTPN11 | |
Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP-1D, PTP-2C, PTP2CNS1, Shp2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11 | |
A synthesized peptide derived from Human SHP-2/PTPN11 [p Tyr542] [UniProt Q06124] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
2E2 | |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Cell Biology, Lipid and Metabolism, Phospho Specific, Signal Transduction, Tyrosine Kinases | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5781 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title