Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHP-2/PTPN11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30348620UL
Description
SHP-2/PTPN11 Polyclonal specifically detects SHP-2/PTPN11 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockout Validated.Specifications
SHP-2/PTPN11 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence, KnockDown | |
Unconjugated | |
BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP-1D, PTP-2C, PTP2CNS1, Shp2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-460 of human SHP-2/PTPN11 (NP_002825.3). | |
20 μL | |
Angiogenesis, Cancer, Cell Biology, Lipid and Metabolism, Phospho Specific, Signal Transduction, Tyrosine Kinases | |
5781 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction