Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHP/NR0B2/Nuclear Receptor SHP Antibody (OTI7B5), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274172C
Description
SHP/NR0B2/Nuclear Receptor SHP Monoclonal specifically detects SHP/NR0B2/Nuclear Receptor SHP in Human samples. It is validated for Western Blot.Specifications
SHP/NR0B2/Nuclear Receptor SHP | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NR0B2 | |
Full length recombinant protein of human SHP/NR0B2/Nuclear Receptor SHP (NP_068804) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot | |
OTI7B5 | |
Western Blot | |
FLJ17090, nuclear receptor subfamily 0 group B member 2, nuclear receptor subfamily 0, group B, member 2, Orphan nuclear receptor SHP, SHPSHP1, Small heterodimer partner | |
Mouse | |
Affinity Purified | |
GPCR | |
8431.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction