Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHPRH Antibody (OTI5G8), DyLight 680, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP274180FR

Catalog No. NBP274180FR


Only null left
Add to Cart

Description

Description

SHPRH Monoclonal specifically detects SHPRH in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

SHPRH
Monoclonal
DyLight 680
PBS with 0.05% Sodium Azide
SHPRH
Recombinant protein fragment corresponding to amino acids 1-100 and 1500-1600 of human SHPRH (NP_775105) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG2b
Western Blot, Flow Cytometry
OTI5G8
Western Blot, Flow Cytometry
2610103K11Rik, bA545I5.2, E3 ubiquitin-protein ligase SHPRH, EC 3.6.4.-, EC 6.3.2.-, FLJ27258, FLJ37625, FLJ90837, histone-linker, PHD and RING finger domain-containing helicase, KIAA2023FLJ45012, MGC134886, SNF2 histone linker PHD RING helicase
Mouse
Affinity Purified
DNA Repair
257218.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.