Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHPRH Monoclonal Antibody (OTI5D4), TrueMAB™, OriGene
SDP

Catalog No. 502542275
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2275 30 μL
50-167-2590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2275 Supplier OriGene Supplier No. TA501446S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SHPRH Monoclonal antibody specifically detects SHPRH in Human samples. It is validated for Western Blot

Specifications

Antigen SHPRH
Applications Western Blot
Classification Monoclonal
Clone OTI5D4
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SHPRH
Gene Accession No. Q149N8
Gene Alias 2610103K11Rik; AA450458; AU024614; bA545I5.2; BC006883; D230017O13Rik; E130018M05; E3 ubiquitin-protein ligase SHPRH; KIAA2023; RING-type E3 ubiquitin transferase SHPRH; Shprh; SNF2 histone linker PHD RING helicase; SNF2 histone linker PHD RING helicase, E3 ubiquitin protein ligase; SNF2, histone-linker, PHD and RING finger domain-containing helicase
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-100 and 1500
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257218
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.