Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SHQ1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534449
Description
A suggested positive control is human heart tissue lysate. PA5-34449 can be used with blocking peptide PEP-1491.
SHQ1 is the human homolog of yeast Shq1p, a protein that acts as an accessory factor in the assembly of H/ACA ribonucleoproteins. SHQ1 has been shown to bind the major H/ACA core protein and the pseudouridine synthase NAP57, but precedes the assembly role of NAF1 and of the other H/ACA core proteins. SHQ1 assists in the assembly of H/ACA-box ribonucleoproteins that function in the processing of ribosomal RNAs, modification of spliceosomal small nuclear RNAs, and stabilization of telomerase.
Specifications
SHQ1 | |
Polyclonal | |
Unconjugated | |
SHQ1 | |
2810403P18Rik; FLJ10539; gene associated with retinoid and interferon-induced mortality 1; GRIM-1; hypothetical protein LOC507486; protein SHQ1 homolog; RGD1310610; Shq1; SHQ1 homolog; SHQ1 homolog (S. cerevisiae); SHQ1, H/ACA ribonucleoprotein assembly factor; Shq1p; zgc:158255 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55164 | |
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6PI26 | |
SHQ1 | |
A 17 amino acid synthetic peptide near the amino terminus of human SHQ1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction