Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHQ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560124
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560124 Supplier Invitrogen™ Supplier No. PA560124
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELKDSPSETV SSLQGPFLEE SSAFLIVDGG VRRNTAIQES DASQGKPLAS SWPLGVSGPL IEELGEQLKT TVQVSEPKGT TAVNRSNIQE RDGCQTPNN Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 32%.

SHQ1 is the human homolog of yeast Shq1p, a protein that acts as an accessory factor in the assembly of H/ACA ribonucleoproteins. SHQ1 has been shown to bind the major H/ACA core protein and the pseudouridine synthase NAP57, but precedes the assembly role of NAF1 and of the other H/ACA core proteins. SHQ1 assists in the assembly of H/ACA-box ribonucleoproteins that function in the processing of ribosomal RNAs, modification of spliceosomal small nuclear RNAs, and stabilization of telomerase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHQ1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SHQ1
Gene Accession No. Q6PI26
Gene Alias 2810403P18Rik; FLJ10539; gene associated with retinoid and interferon-induced mortality 1; GRIM-1; hypothetical protein LOC507486; protein SHQ1 homolog; RGD1310610; Shq1; SHQ1 homolog; SHQ1 homolog (S. cerevisiae); SHQ1, H/ACA ribonucleoprotein assembly factor; Shq1p; zgc:158255
Gene Symbols SHQ1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SHQ1. Recombinant protein control fragment (Product #RP-100146).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55164
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.