Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHROOM1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282005
Description
SHROOM1 Polyclonal specifically detects SHROOM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
SHROOM1 | |
Polyclonal | |
Unconjugated | |
PBS | |
SHROOM1 | |
Rabbit polyclonal SHROOM1 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human SHROOM1. The immunogen is located within the last 50 amino acids of SHROOM1. Amino Acid Squence: CDDLGHHAPSPSPARPPGT | |
Affinity Purified | |
RUO | |
134549 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1-2 ug/mL, ELISA | |
Apical Protein 2, APXL2, KIAA1960, Protein Shroom1, Shroom Family Member 1 | |
Rabbit | |
94 kDa | |
0.1 mg | |
Primary | |
SHROOM1 antibody is human, mouse and rat reactive. At least three isoforms of SHROOM1 are known to exist; this antibody will detect only the two largest isoforms. SHROOM1 antibody is predicted to not cross-react with other SHROOM family members. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction