Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHROOM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562449 Supplier Invitrogen™ Supplier No. PA562449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPLLIQDEDS TRIERVMDNN TTVKMVPIKI VHSESQPEKE SRQSLACPAE PPALPHGLEK DQIKTLSTSE QFYS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.

The protein encoded by this gene shares significant similarities with the apical protein from Xenopus laevis which is implicated in amiloride-sensitive sodium channel activity. This gene is a strong candidate gene for ocular albinism type 1 syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHROOM2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SHROOM2
Gene Accession No. Q13796
Gene Alias 4832440C16; Ab2-404; apical-like protein; APX homolog of Xenopus; Apxl; C630003H05Rik; HSAPXL; liver regeneration-related protein LRRG167; protein Apxl; Protein Shroom2; Shrm2; shroom family member 2; SHROOM2
Gene Symbols SHROOM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SHROOM2. Recombinant protein control fragment (Product #RP-95447).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 357
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.