Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SI-CLP Polyclonal Antibody

SI-CLP Polyclonal Antibody

Supplier:  Thermo Scientific PA595695

Catalog No. PIPA595695


Only null left
Add to Cart

Description

Description

SI-CLP Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SI-CLP
Polyclonal
Unconjugated
CHID1
3110023E09Rik; AI841869; CHID1; chitinase domain containing 1; chitinase domain-containing protein 1; FLJ42707; GL008; MGC3234; PSEC0104; SB139; SICLP; SI-CLP; stabilin-1 interacting chitinase-like protein; stabilin-1-interacting chitinase-like protein
Rabbit
Affinity Chromatography
RUO
68038
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.87 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q922Q9
CHID1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-393 of human CHID1 (NP_0764363).
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.