Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SI-CLP Polyclonal Antibody

Supplier: Thermo Scientific PA5113454
Description
Specifications
| SI-CLP | |
| Polyclonal | |
| Unconjugated | |
| CHID1 | |
| 3110023E09Rik; AI841869; CHID1; chitinase domain containing 1; chitinase domain-containing protein 1; FLJ42707; GL008; MGC3234; PSEC0104; SB139; SICLP; SI-CLP; stabilin-1 interacting chitinase-like protein; stabilin-1-interacting chitinase-like protein | |
| Rabbit | |
| Protein G | |
| RUO | |
| 66005 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9BWS9 | |
| CHID1 | |
| Recombinant Human Chitinase domain-containing protein 1 protein (20-393AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction