Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SI-CLP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558761
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558761 Supplier Invitrogen™ Supplier No. PA558761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KTLLEKSQFS DKPVQDRGLV VTDLKAESVV LEHRSYCSAK ARDRHFAGDV LGYVTPWNSH GYDVTKVFGS KFTQISPV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 90%.

SI-CLP detects human Stabilin-interacting chitinase like protein (SI-CLP), a novel member of the Glyco-18-domain containing protein family. SI-CLP is a secretory protein which is up-regulated in response to interleukin-4 and dexamethasone in activated macrophages. SI-CLP is also expressed by T- and B-lymphocytes, and by cells of monocytic and epithelial origin. SI-CLP is the ligand for stabilin-1, an intracellular sorting receptor which sorts SI-CLP into late endosomes and secretory lysosomes in activated macrophages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SI-CLP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHID1
Gene Accession No. Q9BWS9
Gene Alias 3110023E09Rik; AI841869; CHID1; chitinase domain containing 1; chitinase domain-containing protein 1; FLJ42707; GL008; MGC3234; PSEC0104; SB139; SICLP; SI-CLP; stabilin-1 interacting chitinase-like protein; stabilin-1-interacting chitinase-like protein
Gene Symbols CHID1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SI-CLP. Recombinant protein control fragment (Product #RP-97509).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66005
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.