Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SI-CLP Rat anti-Human, Clone: 1C11, Invitrogen™
Rat Monoclonal Antibody
Supplier: Thermo Scientific MA528222
Description
SI-CLP detects human Stabilin-interacting chitinase like protein (SI-CLP), a novel member of the Glyco-18-domain containing protein family. SI-CLP is a secretory protein which is up-regulated in response to interleukin-4 and dexamethasone in activated macrophages. SI-CLP is also expressed by T- and B-lymphocytes, and by cells of monocytic and epithelial origin. SI-CLP is the ligand for stabilin-1, an intracellular sorting receptor which sorts SI-CLP into late endosomes and secretory lysosomes in activated macrophages.
Specifications
SI-CLP | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q9BWS9 | |
Rat | |
Protein G | |
RUO | |
66005 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
1C11 | |
Unconjugated | |
CHID1 | |
CHID1 | |
SI-CLP fragment corresponding to amino acids 24-73 expressed as a GST fusion protein in E Coli | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction