Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569349
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569349 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569349 Supplier Invitrogen™ Supplier No. PA569349
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 90%; Guinea Pig: 90%; Horse: 79%; Human: 100%; Mouse: 79%; Pig: 93%; Rabbit: 86%; Rat: 90%; Zebrafish: 82%.

This gene encodes a sucrase-isomaltase enzyme that is expressed in the intestinal brush border. The encoded protein is synthesized as a precursor protein that is cleaved by pancreatic proteases into two enzymatic subunits sucrase and isomaltase. These two subunits heterodimerize to form the sucrose-isomaltase complex. This complex is essential for the digestion of dietary carbohydrates including starch, sucrose and isomaltose. Mutations in this gene are the cause of congenital sucrase-isomaltase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SI
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SI
Gene Accession No. P14410
Gene Alias 2010204N08Rik; alpha-glucosidase; c-sis; fd42f04; fd44a09; Isomaltase; maltase-glucoamylase (alpha-glucosidase); maltase-glucoamylase, intestinal; mgam; oligosaccharide alpha-1,6-glucosidase; pdgf protein; PDGF subunit B; PDGF-2; Pdgfb; platelet derived growth factor, B polypeptide; platelet-derived growth factor B chain; Platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); platelet-derived growth factor subunit B; Platelet-derived growth factor, same as Sis (Simian sarcoma viral oncogene homologue, c-sis); pro-sucrase-isomaltase (EC 3.2.1.48-10); SI; SIS; Si-s; SUCIMAL; Sucrase; sucrase isomaltase (alpha-glucosidase); sucrase isomaltase, structural; sucrase-isomaltase; sucrase-isomaltase (alpha-glucosidase); sucrase-isomaltase, intestinal; wu:fd42f04; wu:fd44a09
Gene Symbols SI
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SI.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6476
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.