Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIA7C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563122 Supplier Invitrogen™ Supplier No. PA563122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVNFPLLLNC FGQPGTKWIP FSYTYRRPLR THYGYINVKT QEPLQLDCDL Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 78%.

Involved in the biosynthesis of ganglioside GD1A from GM1B. Transfers CMP-NeuAc with an alpha-2,6-linkage to GalNAc residue on NeuAc-alpha-2,3-Gal-beta-1,3-GalNAc of glycoproteins and glycolipids. ST6GalNAcIII prefers glycolipids to glycoproteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIA7C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ST6GALNAC3
Gene Accession No. Q8NDV1
Gene Alias Alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 3; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase III; GalNAc alpha-2,6-sialyltransferase III; PRO7177; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) C; sialyltransferase 7C; SIAT7C; SIAT7-C; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltran; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 3; ST6 GalNAc alpha-2,6-sialyltransferase 3; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 3; ST6GALNAC III; ST6GALNAC3; ST6GALNACIII; STY; UNQ2787/PRO7177
Gene Symbols ST6GALNAC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SIA7C. Recombinant protein control fragment (Product #RP-101556).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256435
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.