Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIAE Antibody (03), DyLight 650 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306150C
Description
SIAE Monoclonal antibody specifically detects SIAE in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SIAE | |
Monoclonal | |
DyLight 650 | |
AIS6, CSEC, CSE-C, cytosolic sialic acid 9-O-acetylesterase homolog, EC 3.1.1.53, H-Lse, LSE, MGC87009, sialate O-acetylesterase, sialic acid acetylesterase, Sialic acid-specific 9-O-acetylesterase, sialic acid-specific acetylesterase II, YSG2, Ysg2 homolog (mouse) | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SIAE (Q9HAT2-1; Met1-Lys523). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
03 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
54414 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction