Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIAE Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SIAE |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SIAE Polyclonal specifically detects SIAE in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SIAE | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
54414 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ACGVPKQGSIPYDSVTGPSKHSVLWNAMIHPLCNMTLKGVVWYQGESNINYNTDLYNCTFPALIEDWRETFHRGSQGQTERFFPFGLVQLSSDLS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
AIS6, CSEC, CSE-C, cytosolic sialic acid 9-O-acetylesterase homolog, EC 3.1.1.53, H-Lse, LSE, MGC87009, sialate O-acetylesterase, sialic acid acetylesterase, Sialic acid-specific 9-O-acetylesterase, sialic acid-specific acetylesterase II, YSG2, Ysg2 homolog (mouse) | |
SIAE | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title