Learn More
Invitrogen™ SIAE Monoclonal Antibody (03)

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SIAE/sialate O-acetylesterase.
Specifications
Specifications
| Antigen | SIAE |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 03 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | SIAE |
| Gene Accession No. | Q9HAT2 |
| Gene Alias | AIS6; clone 165; CSEC; CSE-C; cytosolic sialic acid 9-O-acetylesterase homolog; H-Lse; LSE; RGD1310431; SIAE; Sialate O-acetylesterase; Sialate O-acetylesterase large subunit; Sialate O-acetylesterase small subunit; sialic acid acetylesterase; sialic acid-specific 9-O-acetylesterase; sialic acid-specific acetylesterase II; yolk sac gene 2; yolk sac protein 2; YSG2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.