Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIAE Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1G5F7, Proteintech
SDP

Catalog No. 501736943 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6943 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6943 Supplier Proteintech Group Inc Supplier No. 665221IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes an enzyme which removes 9-O-acetylation modifications from sialic acids. Mutations in this gene are associated with susceptibility to autoimmune disease 6. Multiple transcript variants encoding different isoforms, found either in the cytosol or in the lysosome, have been found for this gene.

Specifications

Antigen SIAE
Applications Western Blot
Classification Monoclonal
Clone 1G5F7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SIAE
Gene Accession No. P70665, P82450, Q9HAT2
Gene Alias CSE C, H Lse, LSE, SIAE, Sialate O acetylesterase, sialic acid acetylesterase, YSG2
Gene Symbols Siae
Host Species Mouse
Immunogen SIAE Fusion Protein Ag14624
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100521812, 22619, 363045, 54414
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.