Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIAE Polyclonal Antibody
SIAE Polyclonal Antibody
Supplier: Thermo Scientific PA558230
Description
SIAE Polyclonal Antibody for Western Blot, IHC

Specifications
SIAE | |
Polyclonal | |
Unconjugated | |
SIAE | |
AIS6; clone 165; CSEC; CSE-C; cytosolic sialic acid 9-O-acetylesterase homolog; H-Lse; LSE; RGD1310431; SIAE; Sialate O-acetylesterase; Sialate O-acetylesterase large subunit; Sialate O-acetylesterase small subunit; sialic acid acetylesterase; sialic acid-specific 9-O-acetylesterase; sialic acid-specific acetylesterase II; yolk sac gene 2; yolk sac protein 2; YSG2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54414 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HAT2 | |
SIAE | |
Recombinant protein corresponding to Human SIAE. Recombinant protein control fragment (Product #RP-97295). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction