Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIAH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675843
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-843 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-843 Supplier Invitrogen™ Supplier No. PA112863
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for this product is human brain lysate.

This gene encodes a protein that is a member of the seven in absentia homolog family. The protein is an E3 ligase and is involved in ubiquitination and proteasome-mediated degradation of specific proteins. The activity of this ubiquitin ligase has been implicated in the development of certain forms of Parkinson's disease, the regulation of the cellular response to hypoxia and induction of apoptosis. Alternative splicing results in several additional transcript variants, some encoding different isoforms and others that have not been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIAH1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene SIAH1
Gene Accession No. P61092, Q06985, Q8IUQ4
Gene Alias AA960570; AA982064; AI853500; E3 ubiquitin-protein ligase SIAH1; E3 ubiquitin-protein ligase SIAH1A; E3 ubiquitin-protein ligase SIAH1B; FLJ08065; h HUMSIAH; hSIAH1; HUMSIAH; mSiah-1a; RING-type E3 ubiquitin transferase SIAH1; RING-type E3 ubiquitin transferase SIAH1A; RING-type E3 ubiquitin transferase SIAH1B; seven in absentia 1A; seven in absentia 1B; seven in absentia homolog 1; Seven in absentia homolog 1a; Seven in absentia homolog 1b; SIAH; siah E3 ubiquitin protein ligase 1; SIAH1; siah-1; Siah1a; Siah-1a; Siah1b; siah-1b; Sinh1a; Sinh1b
Gene Symbols SIAH1, Siah1a, Siah1b
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human SIAH-1.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110337437, 20437, 20438, 6477
Target Species Human, Mouse
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.