Learn More
Invitrogen™ SIAH1/SIAH2 Monoclonal Antibody (8G7H12)

Description
This is specific for human SIAH 1 and 2 and Drosophila SINA in proliferating cells and stem cells. This does not detect Drosophila SINAH or normal, nondividing cells. Does not react with mouse.
Specifications
Specifications
| Antigen | SIAH1/SIAH2 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 8G7H12 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with 0.1% sodium azide |
| Gene | SIAH2 |
| Gene Accession No. | O43255, P21461, Q06986, Q7ZVG6, Q8IUQ4, Q8R4T2, Q920M9 |
| Gene Alias | AA415433; E3 ubiquitin-protein ligase Siah1; E3 ubiquitin-protein ligase SIAH2; EC 6.3.2; FLJ08065; hSIAH1; hSiah2; HUMSIAH; id:ibd2635; mSiah2; RING-type E3 ubiquitin transferase SIAH1; RING-type E3 ubiquitin transferase SIAH2; seven in absentia 1A; seven in absentia 2; seven in absentia homolog 1; Seven in absentia homolog 2; SIAH; siah E3 ubiquitin protein ligase 1; siah E3 ubiquitin protein ligase 2; Siah1; siah-1; Siah1a; Siah-1a; Siah2; siah-2; Sinh2; zgc:56026 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.