Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIAH1/SIAH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536274
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536274 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536274 Supplier Invitrogen™ Supplier No. PA536274
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 40 kDa. Purity is >95% by SDS-PAGE.

SIAH, the human homologue of the Drosophila seven in absentia (sina) gene, is a tumor suppressor protein that is expressed in intestinal epithelium and activated during apoptosis. Human SIAH proteins are produced as two distinct gene products, SIAH-1, and a slightly larger protein SIAH-2, which share a highly conserved C-terminal sequence and differ in their N-terminal regions. SIAH-1 is a 32 kDa protein that contains an N-terminal RING-finger domain, which is required for proteolysis, and a cystein-rich C-terminal domain, which regulates oligomerization and SIAH binding to target proteins. As a tumor suppressor, SIAH-1 binds DCC (deleted in colorectal cancer) and regulates DCC degradation via the ubiquitin-proteasome pathway. SIAH-1 also binds a Bcl-2 related protein, Bag-1, thereby inhibiting cell growth. The majority of SIAH-1 is localized to the nucleus, however a small percentage is detected in the cytoplasm. This nuclear localization suggests that SIAH proteins may interact with other nuclear matrix proteins and DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIAH1/SIAH2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SIAH2
Gene Accession No. O43255, Q06986, Q8IUQ4, Q8R4T2, Q920M9
Gene Alias AA415433; E3 ubiquitin-protein ligase Siah1; E3 ubiquitin-protein ligase SIAH2; EC 6.3.2; FLJ08065; hSIAH1; hSiah2; HUMSIAH; id:ibd2635; mSiah2; RING-type E3 ubiquitin transferase SIAH1; RING-type E3 ubiquitin transferase SIAH2; seven in absentia 1A; seven in absentia 2; seven in absentia homolog 1; Seven in absentia homolog 2; SIAH; siah E3 ubiquitin protein ligase 1; siah E3 ubiquitin protein ligase 2; Siah1; siah-1; Siah1a; Siah-1a; Siah2; siah-2; Sinh2; zgc:56026
Gene Symbols SIAH1, SIAH2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 178-229 of human Siah-1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110337437, 140593, 140941, 20439, 6477, 6478
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.