Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIAH2 Monoclonal Antibody (24E6H3)
GREENER_CHOICE

Catalog No. PIMA524780
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524780 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524780 Supplier Invitrogen™ Supplier No. MA524780
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This is specific for human SIAH2 and Drosophila SINA in proliferating cells and stem cells. This does not detect Drosophila SINAH or normal, nondividing cells.

SIAH-2 (seven in absentia homolog 2) is an E3 ligase that catalyzes ubiquitination and proteasome-mediated degradation of protein substrates. SIAH-2 encodes a 324 amino acid protein that shares 77% identity with human SIAH-1 and 68% identity with the Drosophila sina (seven in absentia) gene, on which development of the Drosophila R7 photoreceptor is dependent. SIAH-2 targets TRAF2 (which regulates cell responses to stress and cytokines through the regulation of key stress-signaling cascades) for degradation under stress conditions such as hypoxia. It targets HIF-1 alpha prolyl hydroxylase 3 (PHD3) for degradation upon exposure to hypoxic conditions, which coincides with an increase in SIAH-2 transcription. SIAH-2 can decrease TNF-alpha-dependent induction of JNK activity and transcriptional activation of NFkB. SIAH-2 null mice subjected to hypoxia display an impaired respiratory response and reduced levels of hemoglobin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIAH2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 24E6H3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with 0.1% sodium azide
Gene SIAH2
Gene Accession No. O43255, P21461, Q06986
Gene Alias AA415433; E3 ubiquitin-protein ligase SIAH2; EC 6.3.2; hSiah2; mSiah2; RING-type E3 ubiquitin transferase SIAH2; seven in absentia 2; seven in absentia homolog 2; siah E3 ubiquitin protein ligase 2; SIAH2; siah-2; Sinh2
Gene Symbols SIAH2, sina
Host Species Mouse
Immunogen Synthetic peptide made to an N-terminal portion of the drosophila SINA protein (within residues 1-100).
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100622531, 20439, 39884, 6478
Target Species Drosophila, Human, Mouse, Pig
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.