Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sialosyl-Tn Antigen Monoclonal Antibody (STn 219)
GREENER_CHOICE

Catalog No. MA190577
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA190577 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA190577 Supplier Invitrogen™ Supplier No. MA190577
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: Human gastrointestinal tumor, prostate, ovary ca.. This antibody stains colon carcinomas and ovarian mucinous carcinomas producing apical cytoplasmic, apical membrane and luminal staining patterns. It is negative with majority of adult tissue sections.

Sialosyl Tn antigen, also known as CD175s, is expressed in the majority of colon, gastric, breast, and lung carcinoma. The sialosyl Tn has limited expression to being absent in normal colon, pancreas, stomach, ovary, and prostate. This antigen is detected in several adenocarcinomas. It appears to be useful marker for poorly differentiated colon adenocarcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sialosyl-Tn Antigen
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone STn 219
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.05% sodium azide; pH 7.2
Gene Alias CD175s
Host Species Mouse
Immunogen Ovine submaxillary mucin (OSM).
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.