Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SID 7969543
GREENER_CHOICESDP

Catalog No. 344010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-401-0 10 mg
34-405-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 34-401-0 Supplier Tocris Bioscience™ Supplier No. 3440/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective steroidogenic factor-1 (SF-1, NR5A1) inhibitor

Selective steroidogenic factor-1 (SF-1, NR5A1) inhibitor (IC50 values are 0.76, >33 and >33 μM at SF-1, RORα and VP16 respectively). Inhibits SF-1-dependent luciferase expression in HEK 293T cells in vitro (IC50 = 30 nM).

Chemical Identifiers

CAS 868224-64-0
Molecular Formula C24H24N2O7

Specifications

Quantity 10 mg
Formula Weight 452.46
Purity >98%
Molecular Weight (g/mol) 452.463
Chemical Name or Material SID 7969543
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.