Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIDT1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306051C
Description
SIDT1 Polyclonal specifically detects SIDT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SIDT1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
B830021E24Rik, FLJ20174, SID1, SID-1, SID1 transmembrane family member 1, SID1 transmembrane family, member 1 | |
Produced in rabbits immunized with E. coli-derived Human SIDT1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
54847 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction