Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIDT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571390
Description
SIDT1 Polyclonal Antibody for Western Blot
The SIDT1 gene encodes the SID1 transmembrane family member 1 protein, which is involved in RNA transport and may play a role in RNA silencing pathways. SIDT1 has been studied for its role in facilitating the uptake and cellular exchange of double-stranded RNA (dsRNA), a process that is crucial for gene regulation and immune responses. The protein's activity is thought to be involved in RNA interference (RNAi), a mechanism for silencing genes that are complementary to an introduced dsRNA. SIDT1's function is important for maintaining RNA homeostasis and regulating gene expression, impacting various cellular processes and potential therapeutic applications, such as antiviral strategies and cancer treatment. Understanding the role of SIDT1 in RNA transport and the RNAi pathway provides insights into how cells manage genetic and environmental information, which could be pivotal for developing RNA-based therapeutic interventions.
Specifications
| SIDT1 | |
| Polyclonal | |
| Unconjugated | |
| SIDT1 | |
| AW045928; B830021E24Rik; SID1; SID-1; SID1 transmembrane family member 1; SID1 transmembrane family, member 1; SIDT1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54847 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NXL6 | |
| SIDT1 | |
| synthetic peptide directed towards the N terminal region of human SIDT1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction