Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIDT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569064
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%.
The SID1 transmembrane family member (SIDT2) is a highly glycosylated, multipass transmembrane protein that localizes to the lysosomal membrane. SIDT2 is highly expressed in liver, brain and kidney, with no or little expression in skeletal muscles, heart or other tissues. Little is known of the function of SIDT2.
Specifications
SIDT2 | |
Polyclonal | |
Unconjugated | |
SIDT2 | |
B930096O19; BC023957; CGI-40; PSEC0072; RGD1308311; SID1 transmembrane family member 2; SID1 transmembrane family, member 2; Sidt2; UNQ685/PRO1325 | |
Rabbit | |
Affinity chromatography | |
RUO | |
51092 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Surface Plasmon Resonance | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NBJ9 | |
SIDT2 | |
synthetic peptide directed towards the N terminal region of human SIDT2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction